Learn More
National Safety Apparel™ Chemical Resistant FR Lab Coats with Pockets
SDP
Catalog No. 17000302 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-000-302 Medium
17-000-300 X-Small
17-000-301 Small
17-000-303 Large
17-000-304 X-Large
17-000-305 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-000-302 Supplier National Safety Apparel™ Supplier No. LABCRNBMD
Add to Cart
Add to Cart
Navy chemical resistant lab coat made of flame-resistant CR/FR fabric for multi-hazard protection. Pockets feature covered flaps to secure contents.
  • Fabric/Material 4.4 oz. CR/FR Aramid Blend FR Nomex knit wrists keep sleeves in place
  • Bottom side vents for increased mobility
  • Durable hi-temp plastic snaps for easy donning and doffing
  • Side pass-through allows access to pockets underneath the coat
  • Durable accessory loop for safety glasses, pens, badges, etc.
  • Chest pocket with covered flap and pen/pencil slot
  • Lower pockets with covered flaps to secure contents
  • Highly breathable
  • Easy care and laundering
Specifications
Product Type Chemical Resistant Lab Coat
Size Medium
Gender Unisex
Color Navy
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Flame
Style Knee-length
Certifications/Compliance ASTM F1506, NFPA 2112