Learn More
R&D Systems ChemiGlow Chemiluminescence Substrate Kit, 100 mL
SDP
Catalog No. 50921415 Shop All Novus Products
Encompass
Chemiluminescent substrate optimized for CCD digital imaging on FluorChem Systems. HRP-based for Western blots. Includes 100 mL of substrate solution.
Catalog No. 50-921-415 Supplier R&D Systems Supplier No. 601259600
Chemiluminescent substrate optimized for CCD digital imaging on FluorChem Systems. HRP-based for Western blots. Includes 100 mL of substrate solution.