Learn More
Azure Biosystems chemiSOLO Densitometry Kit
SDP
Catalog No. CS1004AZ Shop All Azure Biosystems Products
Change view
Click to view available options
Densitometry Kit
Catalog No.
CS1004AZ Densitometry Kit
1 options
Catalog No. CS1004AZ Supplier Azure Biosystems Supplier No. CS1004
Add to Cart
Add to Cart
Azure Biosystems chemiSOLO Densitometry Kit provides the materials for densitometric validation and analysis using the chemiSOLO imaging system.
  • Allows densitometry analysis to be performed using the Azure Biosystems chemiSOLO imaging system
  • Provides materials for densitometric verification and analysis of visible-stained gels
  • Kit includes: 21-Step Densitometry Calibration Target and chemiSOLO Densitometry Protocol
Specifications
Product Type Chemiluminescence Module