Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Chemistry Hardware Assortment Kit
SDPSCI_ED

Catalog No. S01974A Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S01974A 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S01974A Supplier United Scientific Supplies Supplier No. HRDKIT2C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes laboratory hardware most often used in general chemistry labs.

This set contains one each of:

  • Beaker tong
  • Burette clamp
  • Burner tripod
  • Pinchcock clamp
  • Right angle clamp holder
  • Clay pipe triangle
  • Crucible tong
  • Ring support with 4 in. (10.16cm) clamp
  • Ring stand, 5 x 8 in. (12.7 x 20.32cm) base
  • Test tube rack that holds six tubes
  • Test tube tong
  • Wire gauze, 5 x 5 in. (12.7x 12.7cm)

Specifications

Grade 6 to 12
Product Type Kit, Chemistry Hardware Assortment
Includes Beaker tong, burette clamp, burner tripod, clamp (pinchcock), right angle clamp holder, clay pipe triangle, crucible tong, 4 in. ring support with clamp, ring stand 5 x 8 in. (12.7 x 20.32cm) base, test tube rack (holds 6 tubes), test tube tong and 5 x 5
For Use With (Application) Used in general chemistry labs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.