Learn More
International Enviroguard™ ChemSplash 1 Chemical Splash Coveralls with Serged Seams, Elastic Wrists and Open Ankles
Catalog No. 17184145 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
4X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-184-145 X-Large
17-184-147 Large
17-184-150 2X-Large
17-184-149 3X-Large
17-184-148 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-184-145 Supplier International Enviroguard™ Supplier No. 7012YSXL
Add to Cart
Add to Cart
These durable and disposable chemical coveralls with serged seams and elastic back protect against chemical splash and toxic particles.
  • Durable and disposable chemical coveralls with serged seams and elastic back protect against chemical splash and toxic particles
  • Proprietary, hybrid membrane fabric offers a high moisture vapor transfer rate (MVTR), so you stay cool as you put in a hard day's work
  • A storm flap over the zipper helps prevent hazardous liquids from seeping through otherwise exposed zipper seams
  • For added comfort, seamless shoulder areas help prevent tears in high-movement areas and allow you to move more freely as you work
  • ChemSplash chemical suits also pass ASTM F1670 testing for blood penetration
  • Chemical protective coveralls are designed with serged seams with possible applications including manufacturing, emergency response, environmental cleanup, and more
  • A wider, gusseted crotch gives the wearer more room to move, bend, crouch, and climb without ripping fabric
  • Elastic back stretches with user for more comfort and better tear resistance
  • High moisture vapor transfer rate (MVTR) doesn't trap heat and moisture near the body, so workers stay cooler to prevent heat stress
  • Elastic wrists offer a secure fit that keeps sleeves in place to help prevent skin exposure
Specifications
Product Type Chemical Protective Clothing
Size X-Large
Gender Unisex
Material Laminated Non-woven Fabric
Color Yellow
Disposable Single-use
Waist Style Elastic at Back
Ankle Style Open
Cuff Style Elastic
Neck Style Collared
Hood Type Standard Collar
Closure Type 2-way Front Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Resistance Acid, Caustics, Chemicals, Isopropyl Alcohol, Particle
Latex Free Yes
No. of Pockets None