Learn More
International Enviroguard™ ChemSplash 1 Chemical Splash Coveralls with Taped Seams, Elastic Wrist and Open Ankle
Shop All International Enviroguard Systems ProductsDescription
- Proprietary, hybrid membrane fabric offers a high moisture vapor transfer rate (MVTR), so you stay cool as you put in a hard day's work
- A storm flap over the zipper with pressure-sensitive tape helps prevent hazardous liquids from seeping through exposed zipper seams
- A high-contrast red stripe lets you quickly distinguish this taped-seam garment from coveralls made with serged seams
- For added comfort, seamless shoulders help prevent tears and allow you to move more freely as you work
- Roomy, seamless shoulder area resists tears and provides more protection in key exposure area
- A wider, gusseted crotch gives the wearer more room to move in all directions without ripping fabric
- Elastic back stretches with user for more comfort and better tear resistance
- Storm flap over zipper with pressure-sensitive tape adds an extra layer of protection
- High moisture vapor transfer rate improves wearer comfort
- Elastic wrists offer a secure fit to keep sleeves in place to prevent skin exposure
- Possible applications include manufacturing, emergency response, environmental cleanup, and more
Specifications
Specifications
| Product Type | Chemical Protective Clothing |
| Size | Medium |
| Gender | Unisex |
| Material | Laminated Non-woven Fabric |
| Color | Yellow |
| Disposable | Single-use |
| Waist Style | Elastic at Back |
| Ankle Style | Open |
| Cuff Style | Elastic |
| Neck Style | Collared |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.