Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals CHES, 99%

Catalog No. AAA1804722
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1804722 100 g
AAA1804714 25 g
AAA1804736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1804722 Supplier Thermo Scientific Chemicals Supplier No. A1804722
  • $164.30
    $139.65 / Each of 1

    Save $24.65

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 103-47-9 | C8H17NO3S | 207.288 g/mol

CHES is zwitterionic buffer useful in pH range 8.6 to 10.0.CHES is widely utilized as a buffer which is used for investigations on pH-dependent processes in enzymology. It has been shown to have an unusually high affinity for the iodoacetate binding site of liver alcohol dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
CHES is zwitterionic buffer useful in pH range 8.6 to 10.0.CHES is widely utilized as a buffer which is used for investigations on pH-dependent processes in enzymology. It has been shown to have an unusually high affinity for the iodoacetate binding site of liver alcohol dehydrogenase.

Solubility
Soluble in water.

Notes
Store in cool place. Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-47-9
Molecular Formula C8H17NO3S
Molecular Weight (g/mol) 207.288
MDL Number MFCD00003835
InChI Key MKWKNSIESPFAQN-UHFFFAOYSA-N
PubChem CID 66898
IUPAC Name 2-(cyclohexylamino)ethanesulfonic acid
SMILES C1CCC(CC1)NCCS(=O)(=O)O

Specifications

Melting Point >300°C
Quantity 100 g
Assay Percent Range 0.99
Beilstein 2967601
Merck Index 14,2055
Solubility Information Soluble in water.
Formula Weight 207.29
Percent Purity 99%
Chemical Name or Material CHES
Hazard Category Warning
Hazard Statement GHS H Statement
H319
Precautionary Statement GHS P Statement
P280-P264-P305+P351+P338-P337+P313
EINECSNumber 203-115-6
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.