Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Chetomin, HIF Signaling Inhibitor, Solid, >98% Purity, 1 mg

Catalog No. 502705977
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5977 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5977 Supplier Abcam Supplier No. AB1442221MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Potent HIF signaling inhibitor with >98% purity.

  • Potent inhibitor: HIF signaling inhibitor
  • Molecular weight: 710.9 Da
  • Purity: >98%
  • Form: Solid
  • Biological activity: Reduces CA9 and VEGF expression in hypoxic cells (IC50 = 10 nM, secreted VEGF, Hep38 cells); shows antibacterial, antifungal, and antitumor effects in vivo

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule HIF
Shipping Condition Ambient
Content And Storage Store at -20°C.
Form Solid
Quantity 1 mg
For Use With (Application) HIF signaling inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.