Learn More
Abcam Chetomin, HIF Signaling Inhibitor, Solid, >98% Purity, 5 mg
Description
- Molecular Weight: 710.9 Da
- Purity: >98%
- Potent HIF signaling inhibitor: Reduces CA9 and VEGF expression in hypoxic cells (IC50 = 10 nM, secreted VEGF, Hep38 cells).
- Additional activities: Shows antibacterial, antifungal, and antitumor effects in vivo.
- Form: Solid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Form | Solid |
| Shipping Condition | Ambient |
| Target Molecule | HIF |
| Content And Storage | Store at -20°C. |
| Substrate Properties | Chemical Substrate |
| Quantity | 5 mg |
| For Use With (Application) | HIF signaling inhibition, antibacterial, antifungal, antitumor research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.