Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHI3L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112921
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112921 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112921 Supplier Invitrogen™ Supplier No. PA5112921
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHI3L2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

CHI3L2 may bind glycan structure with high affinity, but not heparin. Has no chitotriosidase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHI3L2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CHI3L2
Gene Accession No. Q15782
Gene Alias CHI3L2; CHIL2; chitinase 3 like 2; chitinase 3-like 2; chitinase-3-like protein 2; Chondrocyte protein 39; YKL39; YKL-39
Gene Symbols CHI3L2
Host Species Rabbit
Immunogen Recombinant Human Chitinase-3-like protein 2 protein (27-390AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1117
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.