Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ChIC/Cut & Run Kit (pG-MN)

Catalog No. CHR101MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
12 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CHR101MI 12 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CHR101MI Supplier MilliporeSigma™ Supplier No. CHR101
Add to Cart
Add to Cart

ChIC/Cut & Run Kit (pA-MN) low signal-to-noise ratio and less starting sample compared to ChIP

  • Benefits:
    • Extremely low signal-to-noise ratio than cross-linking ChIP-seq
    • Cross-linking eliminated (no sonicators or spin steps needed)
    • Lower number of starting cells (minimum 100 cell for histone modification and 1,000 cells for a transcription factor)
    • Shortened procedure time- from cells to purified DNA completed within one day (min. 3 hours)
    • No special equipment needed
  • Includes:
    • Concanavalin A-coated magnetic beads
    • Binding Buffer
    • Wash Buffer
    • Spermidine
    • 0.5M EDTA
    • Rinse Buffer
    • Incubation buffer
    • Stop Solution
    • 2% Digitonin containing DMSO
    • RNase A
    • Spike-in DNA
    • Precipitate Enhancer
    • Proteinase K Solution
    • Protease Inhibitor Cocktail III, Animal Free **Contains DMSO
    • Rabbit IgG Purified
    • Anti-H3K27me3 rabbit monoclonal antibody
    • Protein G- micrococcal nuclease (pG-MN) fusion protein 
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store ChIC box 1 (at 2°C to 8°C and ChIC box 2 (at -20 °C))
For Use With (Application) Low Signal-to-Noise Ratio, Less Starting Sample
Quantity 12 Reactions

For research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.