Learn More
Invitrogen™ Chicken Anemia Virus VP2 Polyclonal Antibody

Description
This antibody has been tested in direct-ELISA.
Specifications
Specifications
| Antigen | Chicken Anemia Virus VP2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 4.77 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene Alias | VP2 |
| Host Species | Rabbit |
| Immunogen | Recombinant Chicken anemia virus Dual specificity protein phosphatase VP2 protein (1-216aa). |
| Purification Method | Protein G |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.