Learn More
Kingfisher Biotech Inc Chicken ANG-4 Protein
SDP
Supplier:  Kingfisher Biotech Inc RP2154C025
Encompass_Preferred
The Chicken ANG-4 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Chicken ANG-4 applications are for cell culture, ELISA standard, and Western Blot Control. Chicken ANG-4 yeast-derived recombinant protein can be purchased in multiple sizes. Chicken ANG-4 Specifications: (Molecular Weight: 13.8 kDa) (Amino Acid Sequence: QNEGYEKFLR QHYDAKPKGR DDRYCESMMK ERKLTSPCKD VNTFIHGTKK NIRAICGKKG SPYGENFRIS NSPFQITTCT HSGASPRPPC GYRAFKDFRY IVIACEDGWP VHFDESFISP (120)) (Gene ID: 100302572). For research use only. Made in the USA
Catalog No. 50-296-4565
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.