Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chicken anti-Rabbit IgG (H+L) Secondary Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NBP175571 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP175571 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP175571 Supplier R&D Systems Supplier No. NBP175571

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Chicken Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunomicroscopy.

Specifications

Antigen IgG (H+L)
Applications Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunomicroscopy
Classification Polyclonal
Concentration LYOPH
Conjugate DyLight 594
Dilution Flow Cytometry 1:20 - 1:2000, Immunocytochemistry/Immunofluorescence, Immunomicroscopy 1:20 - 1:2000
Host Species Chicken
Immunogen Purified Rabbit IgG (H&L)
Purification Method Affinity purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity Based on IEP, this antibody reacts with heavy gamma chains on rabbit IgG and light chains on all rabbit immunoglobulins
Reconstitution Rehydrate with 1.1 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily.
Target Species Rabbit
Content And Storage Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.