Learn More
Kingfisher Biotech Inc Chicken CD40 Ligand (CD40LG) Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP0942C025
Encompass_Preferred
The Chicken CD40 Ligand Ligand yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Chicken CD40 Ligand applications are for cell culture, ELISA standard, and Western Blot Control. The Chicken CD40 Ligand yeast-derived recombinant protein can be purchased in multiple sizes. Chicken CD40 Ligand Specifications: (Molecular Weight: 18.2 kDa) (Amino Acid Sequence: MHRGHEHPHL KSRNETSVAE EKRQPIATHL AGVKSNTTVR VLKWMTTSYA PTSSLISYHE GKLKVEKAGL YYIYSQVSFC TKAAASAPFT LYIYLYLPME EDRLLMKGLD THSTSTALCE LQSIREGGVF ELRQGDMVFV NVTDSTAVNV NPGNTYFGMF KL (162)) (Gene ID: 395485). For research use only. Made in the USA
Catalog No. 50-241-3815
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.