Learn More
Life Diagnostics Inc Chicken Ex-FABP ELISA kit
SDP
Supplier:  Life Diagnostics Inc EXFABP5
Encompass_Preferred
The assay uses two chicken Ex-FABP antibodies developed at Life Diagnostics; one for solid-phase immobilization (microtiter wells) and one, conjugated to horseradish peroxidase (HRP), for detection. Standards and diluted samples are incubated in the microtiter wells with HRP-conjugate for one hour. This results in Ex-FABP molecules being sandwiched between the immobilization and detection antibodies. The wells are washed and TMB is added and incubated for 20 minutes. If Ex-FABP is present a blue color develops. Color development is stopped by the addition of Stop solution, changing the color to yellow. Absorbance is measured at 450 nm. The concentration of Ex-FABP is proportional to absorbance and is derived from a standard curve.
Catalog No. NC1793544
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.