Learn More
Kingfisher Biotech Inc Chicken GM-CSF Biotinylated Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RPB1873C010
Encompass
The Chicken GM-CSF Biotinylated yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Chicken GM-CSF Biotinylated applications are for cell culture. Chicken GM-CSF Biotinylated yeast-derived recombinant protein can be purchased in multiple sizes. Chicken GM-CSF Biotinylated Specifications: (Molecular Weight: 14.2 kDa) (Amino Acid Sequence: TTTYSCCYKV YTILEEITSH LESTAATAGL SSVPMDIRDK TCLRNNLKTF IESLKTNGTE EESGIVFQLN RVHECERLFS NITPTPQVPD KECRTAQVSR EKFKEALKTF FIYLSDVLPE EKDCI (125)) (Gene ID: 416325). For research use only. Made in the USA
Catalog No. 50-253-2337
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.