Learn More
Kingfisher Biotech Inc Chicken IFN alpha Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP0410C500
Encompass
The Chicken IFN alpha yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Chicken IFN alpha applications are for cell culture, ELISA standard, and Western Blot Control. The Chicken IFN alpha yeast-derived recombinant protein can be purchased in multiple sizes. Chicken IFN alpha Specifications: (Molecular Weight: 19.0 kDa) (Amino Acid Sequence: CNHLRPQDAT FSHDSLQLLR DMAPTLPQLC PQHNASCSFN DTILDTSNTR QADKTTHDIL QHLFKILSSP STPAHWNDSQ RQSLLNRIHR YTQHLEQCLD SSDTRSRTRW PRNLHLTIKK HFSCLHTFLQ DNDYSACAWE HVRLQARAWF LHIHNLTGNT RT (162)) (Gene ID: 396398). For research use only. Made in the USA
Catalog No. 50-241-3548
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.