Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Chicken IgM Native Protein
SDP

Catalog No. NBP196785 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP196785 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP196785 Supplier Novus Biologicals™ Supplier No. NBP196785
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

Native Chicken IgM. Source: Serum The Chicken IgM Native Protein has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, SDS-Page.

Specifications

For Use With (Application) ELISA, Immunohistochemistry, SDS-PAGE, Western Blot
Formulation 0.1 M Tris Chloride, 0.5M Sodium Chloride, pH 8.0.
Name Chicken IgM Protein
Quantity 1 mg
Storage Requirements Store at 4°C short term. For extended storage, add an equal volume of glycerol, aliquot and store at -20°C or below. Avoid repeated freeze-thaw cycles.
Cross Reactivity Chicken
Purity or Quality Grade Multi-step
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.