Learn More
Kingfisher Biotech Inc Chicken IL-12 p40 Biotinylated Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RPB1864C010
Encompass
The Chicken IL-12/IL-23 p40 Biotinylated yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Chicken IL-12/IL-23 p40 Biotinylated applications are for cell culture. Chicken IL-12/IL-23 p40 Biotinylated yeast-derived recombinant protein can be purchased in multiple sizes. Chicken IL-12/IL-23 p40 Biotinylated Specifications: (Molecular Weight: 34.1 kDa) (Amino Acid Sequence: AQWKLRENVY VIESEWNDET PAKKVKLTCD TSDEALPVYW KKGTELKGTG KTLTTEVKEF PDAGNYTCLS AKTHEIISYS FFLITKVDSN GQMIRSILKS YKEPSKTFLK CEAKNYSGIF TCSWMTENES PSVKFTIRSL KGSQGDVTCS SPVARTDKSV TEYTAQCQKE NYCPFAEEHQ PTEMFLEVID EVEYENYTSS FFIRDIIKPD PPQCQYASTN GTVTWTYPKT WSTPKSYFPL TFRVKVESTK KYKSKVYDAD EQSIQIPKTG PKDKISVQAR DRYYNSSWSE WSTLCR (296)) (Gene ID: 404671). For research use only. Made in the USA
Catalog No. 50-253-2340
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.