Learn More
Kingfisher Biotech Inc Chicken IL-17B Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1892C005
Encompass
The Chicken IL-17B yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Chicken IL-17B applications are for cell culture, ELISA standard, and Western Blot Control. Chicken IL-17B yeast-derived recombinant protein can be purchased in multiple sizes. Chicken IL-17B Specifications: (Molecular Weight: 17.2 kDa) (Amino Acid Sequence: GPSRLPTAAS ALAWAPDDPY TPMEDYERSI QDMVHQLRNG SEPGDTKCQV NLRLWRSNRR SLSPWAYSIN HDATRIPADI PEARCLCTGC INPFTMQEDR TMASIPIYSR LPVRRLLCQA PGKVGHKASG RKKCHKKYQT VMETIAVGCT CIF (153)) (Gene ID: 427620). For research use only. Made in the USA
Catalog No. 50-253-2630
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.