Learn More
Kingfisher Biotech Inc Chicken M-CSF Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1335C500
Encompass_Preferred
The Chicken M-CSF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Chicken M-CSF applications are for cell culture, ELISA standard, and Western Blot Control. Chicken M-CSF yeast-derived recombinant protein can be purchased in multiple sizes. Chicken M-CSF Specifications: (Molecular Weight: 27.2 kDa) (Amino Acid Sequence: NSYCQQIITE RHLDHLQELA DTQMQQPGTV SFRFISKMRL SDSVCYVKAA FPLLGTILNR TTFKENSTNA NKMKTVRKMY ENIDEDVDPC IRDEDDEEHA LSEMCFEEFT TSPYEMLVLV RQFFQDIKQL LQNKETFEKD CSQVYRSACA GPRQHSSSPE QRRKEPPAAS GGHRELLVYV TVASVVAVLL AMGGLLFYKY KSKVLQRGAA LKEGGCDPEE PESRALQGAQ GCAELETQEL (240)) (Gene ID: 100499189). For research use only. Made in the USA
Catalog No. 50-253-3358
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.