Learn More
American Research Products Inc Chicken oVotransferrin OT EL
Supplier:  American Research Products Inc CSBEL027592CH
Encompass
Chicken oVotransferrin (OT) ELISA kit from CUSABIO Detection Range 0 156 g/mL-20 g/mL Sensitivity 0 156 g/mL Sample serum plasma egg white Method Competitive
Catalog No. 50-003-27496
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.