Learn More
Kingfisher Biotech Inc Chicken Prolactin Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP2031C025
Encompass_Preferred
The Chicken Prolactin yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Chicken Prolactin applications are for cell culture, ELISA standard, and Western Blot Control. Chicken Prolactin yeast-derived recombinant protein can be purchased in multiple sizes. Chicken Prolactin Specifications: (Molecular Weight: 22.7 kDa) (Amino Acid Sequence: LPICPIGSVN CQVSLGELFD RAVKLSHYIH YLSSEIFNEF DERYAQGRGF ITKAVNGCHT SSLTTPEDKE QAQQIHHEDL LNLVVGVLRS WNDPLIHLAS EVQRIKEAPD TILWKAVEIE EQNKRLLEGM EKIVGRVHSG DAGNEIYSHW DGLPSLQLAD EDSRLFAFYN LLHCLRRDSH KIDNYLKVLK CRLIHDSN (198)) (Gene ID: 396453). For research use only. Made in the USA
Catalog No. 50-253-2509
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.