Learn More
Kingfisher Biotech Inc Chicken SCF Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1555C100
Encompass_Preferred
The Chicken SCF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Chicken SCF applications are for cell culture, ELISA standard, and Western Blot Control. Chicken SCF yeast-derived recombinant protein can be purchased in multiple sizes. Chicken SCF Specifications: (Molecular Weight: 22.5 kDa) (Amino Acid Sequence: QSSCGNPVTD DVNDIAKLVG NLPNDYLITL KYVPKMDSLP NHCWLHLMVP EFSRSLHNLL QKFSDISDMS DVLSNYSIIN NLTRIINDLM ACLAFDKNKD FIKENGHLYE EDRFIPENFF RLFNSTIEVY KEFADSLDKN DCIMPSTVET PENDSRVAVT KTISFPPVAA SSLRNDSIGS NTSSNSNKEA LGFISSSSLQ (200)) (Gene ID: 396028). For research use only. Made in the USA
Catalog No. 50-253-3147
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.