Learn More
American Research Products Inc Chicken Testosterone T ELISA
Supplier:  American Research Products Inc CSBE12797C
Encompass
Chicken Testosterone (T) ELISA Kit from CUSABIO Detection Range 0 1 ng/mL-20 ng/mL Sensitivity 0 05 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive
Catalog No. 50-003-29191