Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Chikungunya Virus Capsid Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143450 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143450 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143450 Supplier Invitrogen™ Supplier No. PA5143450
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

Chikungunya virus (CHIKV) belongs to the alphavirus family, the members of which have enveloped icosahedral capsids The capsid protein, an exclusive requirement for NC formation of infectious wild-type (WT) CHIKV particles, is dispensable for the assembly of infectious delta C-CHIKV particles [PMID: 31092567].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chikungunya Virus Capsid
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% Proclin 300; pH 7
Gene Alias CHIKV capsid
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of Chikungunya virus Capsid. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.