Learn More
Invitrogen™ Chikungunya Virus E1 Recombinant Rabbit Monoclonal Antibody (HL2069)
Shop All Thermo Scientific Products

Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Specifications
Specifications
| Antigen | Chikungunya Virus E1 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | HL2069 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene Alias | Chikungunya Virus |
| Host Species | Rabbit |
| Immunogen | Recombinant fragment of Chikungunya virus E1. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.