Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Chikungunya Virus NSP3 Recombinant Rabbit Monoclonal Antibody (HL2479)
GREENER_CHOICE

Catalog No. FEMA550547
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550547 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550547 Supplier Invitrogen™ Supplier No. MA550547
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Chikungunya Virus NSP3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

Chikungunya virus (CHIKV) is a re-emerging Alphavirus causing fever, joint pain, skin rash, arthralgia, and occasionally death The non-structural protein 3 (nsP3) plays an essential role in RNA replication The nsP3 alphavirus unique domain (AUD) is a multi-functional domain It is not only a critical determinant of both cell and species-specificity, but also plays roles in virus genome replication and assembly [PMID: 30668592].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chikungunya Virus NSP3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2479
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias CHIKV NSP3; NsP3 protein
Host Species Rabbit
Immunogen Recombinant fragment of Chikungunya virus nsP3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.