Learn More
Ergodyne™ Chill-Its™ 5157 Premium Cargo Hydration Pack
Catalog No. 19819784 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Black
Hi-Vis Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-784 Black
19-819-785 Hi-Vis Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-819-784 Supplier Ergodyne™ Supplier No. 13163
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Premium Cargo Hydration Pack is designed to keep workers organized and hydrated throughout the day.
  • Durable 600D water-resistant rip stop polyester shell
  • Vented shoulder straps and back for maximum breathability
  • 8 mm EVA foam padding on shoulder straps for added comfort
  • Five zippered compartments, multiple d-rings and nylon daisy chain webbing for gear storage
  • Insulated pack and tube keeps water cool
  • 100% anti-microbial bite valve with cover to protect against contaminants
  • Breakaway shoulder straps for added safety
  • DualCap bladder featuring smaller (60 mm) cap nestled into larger (80 mm) cap for easy water and ice filling
Specifications
Material 600D Polyester
Color Black
Closure Type DualCap
Capacity 3 L