Learn More
Description
- FDA-approved food grade silicone body
- Elastic strap for secure grip
- Valve cover keeps debris off valve
- Valve self-seals, eliminating drips
- Easy to apply
- Fits with all Ergodyne™ hydration packs
Specifications
Specifications
| Material | Silicone |
| Color | Blue |
| For Use With (Application) | Pumping a hydration pack |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.