Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6225 Premium FR Phase Change Cooling Vest

Catalog No. 19819683 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Orange
Tan
Size:
Large/X-Large
Small/Medium
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-819-683 Orange Small/Medium
19-819-676 Tan Small/Medium
19-819-682 Tan Large/X-Large
19-819-684 Orange Large/X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-819-683 Supplier Ergodyne™ Supplier No. 12205
Add to Cart
Add to Cart

Chill-Its™ 6215 Premium Phase Change Cooling Vest keeps workers cool and safe when combined with a cooling pack.

  • Wash by hand with a mild detergent and warm water
  • Flame resistant
  • Made with modacrylic cotton blend FR fabric
  • Adjustable hook and loop closures on both shoulders and waist
  • Extended vest surface area enhances cooling power
  • Designed for more visibility on busy streets or job sites
  • Two elastic pen loops on chest

Specifications

Material Modacrylic Cotton Blend FR Fabric
Product Type Cooling Vest
Size Small/Medium
Length (Metric) 81 to 102 cm
Length (English) 32 to 40 in.
Color Orange
Certifications/Compliance ASTM F1506 performance requirements (ATPV rating of 7.1 cal/cm2); Classified to NFPA 70E (CAT1)
For Use With (Application) Fire Rescue, EMT, Construction
Quantity 6/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.