Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6240 Phase Change Cooling Vest Elastic Extenders

Catalog No. 19819685 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Black
Orange
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-685 Black
19-819-686 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-819-685 Supplier Ergodyne™ Supplier No. 12208

Chill-Its™ 6240 Phase Change Cooling Vest Extenders provide 3.5 in. (9 cm) of extension when used with the Chill-Its™ Phase Change Vest.

  • Comfortable stretch
  • Made of 100% elastic with hook and loop extenders
  • For use with cooling vests

Specifications

Material Cotton
Length (Metric) 9 cm
Length (English) 3.5 in.
Color Black
For Use With (Application) Fire, Rescue, EMT, Construction, Utilities
Quantity 6/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.