Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6485 Multi-Band

Catalog No. 19819619 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Black, White
Blue, White
Hi-Vis Orange
Hi-Vis Yellow
Realtree Xtra™
Red, Black, White
Red, Orange, Black
Red, White, Blue
White
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-619 Hi-Vis Orange
19-819-609 Red, White, Blue
19-819-617 Blue, White
19-819-618 Hi-Vis Yellow
19-819-621 Black, White
19-819-628 Red, Black, White
19-819-607 White
19-819-620 Red, Orange, Black
19-819-627 Realtree Xtra™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-819-619 Supplier Ergodyne™ Supplier No. 42109
Add to Cart
Add to Cart

Chill-Its™ 6485 Multi-Band features supremely versatile performance fabric with a multitude of uses and ways to wear.

  • Multiple style and usage configurations
  • Hi-tech moisture management performance fabric dries fast
  • Multi-climate usage
  • Blocks wind, sun and dust
  • Stretchable, comfortable, and seam-free
  • Machine washable
  • Comes in the following patterns: Red Western, Navy Western, Hi-Vis Yellow, Hi-Vis Orange, Stars and Stripes, Flames, Skulls, Be Tenacious, Realtree™
  • One size

Specifications

Material Hi-tech Moisture Management Performance Fabric
Size Universal
Color Hi-Vis Orange
For Use With (Application) Construction, Landscaping, Maintenance
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.