Learn More
Description
- Multiple style and usage configurations
- Hi-tech moisture management performance fabric dries fast
- Multi-climate usage
- Blocks wind, sun and dust
- Stretchable, comfortable, and seam-free
- Machine washable
- Comes in the following patterns: Red Western, Navy Western, Hi-Vis Yellow, Hi-Vis Orange, Stars and Stripes, Flames, Skulls, Be Tenacious, Realtree™
- One size
Specifications
Specifications
| Material | Hi-tech Moisture Management Performance Fabric |
| Size | Universal |
| Color | Black, White |
| For Use With (Application) | Construction, Landscaping, Maintenance |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.