Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6602 Evaporative Cooling Towel

Catalog No. 19819644 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Gray
Hi-Vis Lime
Orange
Pink
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-644 Gray
19-819-647 Hi-Vis Lime
19-819-648 Orange
19-819-649 Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-819-644 Supplier Ergodyne™ Supplier No. 12438
Add to Cart
Add to Cart

Chill-Its™ 6602 Evaporative Cooling Towel uses a PVA material to absorb water to allow slow evaporation for instant, long-lasting cooling relief.

  • Run under water for one minute or until saturated, wring out excess water for drip free cooling
  • Can last for up to 4 hours on a single soak. Simply re-wet to increase cooling duration
  • Cooling PVA material absorbs water and creates a cooling sensation as it evaporates
  • Air dry until material is stiff, re-wet until soft
  • Machine washable with mild detergent, air dry only
  • Reduces the effects of heat exhaustion and fights fatigue

Specifications

Material PVA
Product Type Cooling Towel
Color Gray
Dimensions (L x W) 13 x 29.5 in. (33 x 75 cm)
For Use With (Application) Construction, Agriculture, Landscaping
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.