Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6602MF Evaporative Microfiber Cooling Towel

Catalog No. 19819659 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Black
Blue
Lime
Orange
Pink
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-659 Black
19-819-653 Blue
19-819-657 Orange
19-819-658 Pink
19-819-660 Lime
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-819-659 Supplier Ergodyne™ Supplier No. 12668

Chill-Its™ 6602MF Microfiber Cooling Towel is designed to create a long-lasting cool feeling while absorbing and holding water.

  • Run under water for one minute or until saturated, wring out excess water for drip-free cooling sensation as it evaporates
  • Made with lightweight, absorbent microfiber material
  • Stays soft either wet or dry
  • ChillX™ microfiber material hold 30% more water and cools 30% longer
  • UPF 50 provides sun protection from harmful UV rays
  • Machine washable with mild detergent, air dry only
  • Reduces the effects of heat exhaustion and fights fatigue

Specifications

Material Microfiber
Color Black
Dimensions (L x W) 9.8 x 40.9 in. (24.9 x 103.9 cm)
Certifications/Compliance UPF 50
For Use With (Application) Construction,Freight/Baggage,Drilling/Mining,Iron/Steel Work,Agriculture,Solar,Landscaping/Grounds
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.