Learn More
Description
- Hi Cool™ moisture management performance fabric
- Absorbent terry headband
- Elasticized back for a great fit
- Available in the following patterns: Stars and Stripes, Camouflage, Red Western, Navy Western, Blue, Lime, Flames, and Skulls
Specifications
Specifications
| Material | Hi Cool™ Moisture Management Performance Fabric |
| Product Type | Headband |
| Color | Gray, Black, White |
| For Use With (Application) | Construction, Maintenance, Landscaping |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.