Learn More
Description
Technical fabric moves moisture away from body for fast evaporation.
- Made of Hi Cool™ Moisture Management performance fabric
- Elasticized back for a great fit
- Machine washable
Specifications
Specifications
| Material | Fabric |
| Product Type | Absorptive Cooling Headband |
| Color | Navy Western |
| For Use With (Application) | Construction, trades, maintenance, landscaping/grounds, assembly/fabrication, material handling, freight/baggage, warehousing/distribution, Iron/Steel fabrication, jungle love |
| Includes | Elasticized Back |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.