Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6615 High-Performance Dew Rag

Catalog No. 19819531 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Black
Gray, Black, White
Lime
Realtree Xtra™
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-531 Black
19-819-524 Lime
19-819-540 Gray, Black, White
19-819-523 Realtree Xtra™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-819-531 Supplier Ergodyne™ Supplier No. 12487
This item is not returnable. View return policy

High-Performance Dew Rag features technical fabric to keep away moisture and an elastic band for a secure fit.

  • Hi Cool™ moisture management performance fabric
  • Comfortable to wear under hats, helmets, or alone
  • Absorbent terry headband
  • Elastic band for a great fit
  • Available in a variety of patterns and colors like black, lime, skulls, and Realtree Xtra™

Specifications

Product Type Dew Rag
Color Black
For Use With (Application) Construction, Maintenance, Landscaping
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.