Learn More
Description
- Hi Cool™ Moisture Management performance fabric
- Comfortable to wear under hats, helmets, or alone
- Absorbent terry headband
- Elastic band
Specifications
Specifications
| Material | Fabric |
| Product Type | High Performance Dew Rag |
| Color | Navy Western |
| For Use With (Application) | Construction, trades, maintenance, landscaping/grounds, assembly/fabrication, material handling, freight/baggage, warehousing/distribution, Iron/Steel fabrication, alligator wrangler |
| Includes | Elastic Band |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.