Learn More
Description
- Hi Cool™ moisture management performance fabric
- Comfortable to wear under hats, helmets or alone
- Wide, absorbent terrycloth band
- One size fits most
- Available in a varisty of colors and patterns like blue, black, skulls, and navy western
Specifications
Specifications
| Material | Hi Cool™ Moisture Management Performance Fabric |
| Size | Universal |
| Color | Blue, White |
| For Use With (Application) | Construction, Maintenance, Landscaping, Distribution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.