Learn More
Description
- 100% Polyester fabric
- Universal design
- Elastic inner rim to fit most hard hats
- Designed to block sun and keep workers comfortable in outdoor environments
Specifications
Specifications
| Color | Lime |
| For Use With (Application) | Construction, Service, Landscaping, Material Handling |
| Material | Polyester |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.