Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6665 Evaporative Cooling Vest

Catalog No. 19910206 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Gray
Lime
Size:
Medium
Large
X-Large
2X-Large
3X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-910-206 Gray Medium
19-910-202 Lime Medium
19-910-203 Lime Large
19-910-204 Lime X-Large
19-910-205 Lime 2X-Large
19-910-207 Gray Large
19-910-208 Gray X-Large
19-910-209 Gray 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-910-206 Supplier Ergodyne™ Supplier No. 12543
Add to Cart
Add to Cart

Evaporative Cooling Vest provides cooling relief and comfort once soaked in cold water.

  • V-neck with zipper closure
  • Quilted nylon outer layer
  • Activated, polymer-embedded fabric inside
  • Water-repellent liner and black cotton-poly elastic trim
  • Lightweight, durable, and washable
  • Soak in water 2-5 minutes to activate
  • Remains hydrated for up to 4 hours
  • Mesh side panels offer improved ventilation and stretch

Specifications

Material Nylon
Product Type Vest
Size Medium
Color Gray
For Use With (Application) Construction, Maintenance, Landscaping, Material Handling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.