Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6686 Dry Evaporative Cooling Hat

Catalog No. 19819606 Shop All Ergodyne Products
Change view
Click to view available options
Color: Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-819-606 Color: Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-819-606 Supplier Ergodyne™ Supplier No. 12680

Chill-Its™ 6686 Dry Evaporative Cooling Hat provides immediate and long-lasting relief from the heat.

  • Dry technology keeps wearer completely dry and comfortable during the entire evaporative cooling process
  • Up to 3 days of dry cooling
  • Fill hat with 3 oz. (89 mL) of water to activate
  • Made of industrial-grade quality nylon
  • Air holes in cap provide continuous ventilation
  • One size fits most
  • Machine washable

Specifications

Size Universal
Color Gray
For Use With (Application) Construction, Maintenance, Landscaping, Material Handling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.