Learn More
Description
- Performance knit fabric is soft, stretchable and durable
- Instant cooling when wet that can last for hours
- Wicks moisture when dry to absorb sweat from the body
- UPF 50+ Protection
- Protection from light abrasions
- Reflective accents
- Anti-odor treatment
- Machine washable
- Chemical-free cooling
Specifications
Specifications
| Material | Performance Knit Fabric |
| Product Type | Arm Sleeves |
| Size | Large |
| Color | Black |
| For Use With (Application) | Agriculture, Construction, Utilities, Transportation, Sports |
| Quantity | 1/Ea. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.