Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6700 Evaporative Cooling Tie Bandana

Catalog No. 19819475 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Black
Camouflage
Gray, Black, White
Lime
Red, Orange, Black
Red, White, Blue
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-475 Camouflage
19-819-473 Lime
19-819-474 Red, White, Blue
19-819-494 Gray, Black, White
19-819-505 Red, Orange, Black
19-819-496 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-819-475 Supplier Ergodyne™ Supplier No. 12304

Evaporative Cooling Bandana features activated polymers that bring cooling power to a variety of application.

  • Activate by soaking in water for 2-5 minutes
  • Acrylic polymer crystals
  • Remains hydrated for up to 4 hours
  • Reusable
  • Available in a variety of colors and patterns like camo, stars and stripes, skulls, and flames

Specifications

Product Type Bandana
Color Camouflage
Closure Type Tie
For Use With (Application) Construction, Maintenance, Landscaping, Material Handling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.