Learn More
Ergodyne™ Chill-Its™ 6700CT Evaporative Cooling Tie Bandana with Cooling Towel
Shop All Ergodyne ProductsDescription
- Advanced PVA material activates easily and quickly when run under water for one minute
- Reusable
- Super-evaporative cooling process helps keep you cool
- Lightweight and low profile
- Available in a variety of colors and patterns like blue, hi-vis lime, red western, blue western, stars and stripes, flames, camouflage, and skulls
Specifications
Specifications
| Material | PVA |
| Product Type | Bandana |
| Color | Camouflage |
| Closure Type | Tie |
| For Use With (Application) | Construction, Maintenance, Landscaping, Material Handling |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.