Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6700CT Evaporative Cooling Tie Bandana with Cooling Towel

Catalog No. 19819548 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Black
Blue
Blue, White
Camouflage
Gray, Black, White
Lime
Red, Orange, Black
Red, White
Red, White, Blue
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-548 Red, White
19-819-546 Red, White, Blue
19-819-547 Camouflage
19-819-550 Blue, White
19-819-551 Black
19-819-552 Lime
19-819-553 Blue
19-819-554 Red, Orange, Black
19-819-555 Gray, Black, White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-819-548 Supplier Ergodyne™ Supplier No. 12563
Add to Cart
Add to Cart

Evaporative Cooling Bandana with Cooling Towel features advanced PVA material, which keeps you cool without any bulk.

  • Advanced PVA material activates easily and quickly when run under water for one minute
  • Reusable
  • Super-evaporative cooling process helps keep you cool
  • Lightweight and low profile
  • Available in a variety of colors and patterns like blue, hi-vis lime, red western, blue western, stars and stripes, flames, camouflage, and skulls

Specifications

Material PVA
Product Type Bandana
Color Red, White
Closure Type Tie
For Use With (Application) Construction, Maintenance, Landscaping, Material Handling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.