Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6705CT Evaporative Hook and Loop Bandana with Cooling Towel

Catalog No. 19819557 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Black
Blue
Blue, White
Camouflage
Gray, Black, White
Lime
Red, Orange, Black
Red, White
Red, White, Blue
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-557 Camouflage
19-819-559 Blue, White
19-819-560 Lime
19-819-556 Red, White, Blue
19-819-558 Red, White
19-819-561 Blue
19-819-562 Red, Orange, Black
19-819-563 Gray, Black, White
19-819-564 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-819-557 Supplier Ergodyne™ Supplier No. 12572

Chill-Its™ 6705CT Evaporative Hook and Loop Bandana with Cooling Towel features advanced PVA material that keeps you cool without the bulk.

  • Advanced PVA material activates easily and quickly when run under water for one minute
  • Reusable
  • Super-evaporative cooling process helps keep you cool
  • Breakaway hook and loop closure
  • Available in a variety of colors and patterns like red western, blue western, camo, stars and striped, black, lime, blue, skulls, and flames

Specifications

Material PVA
Product Type Bandana
Color Camouflage
Closure Type Hook and Loop
For Use With (Application) Construction, Maintenance, Construction, Landscaping, Material Handling
Includes Cooling Towel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.